Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human TGF-beta 3, Animal-Free Protein

Catalog No. QK0540500
Click to view available options
Quantity:
50 μg
500 μg
This item is not returnable. View return policy
This item is not returnable. View return policy
No significant difference between EC50 of reference and test lots

Specifications

Accession Number P10600.1
For Use With (Application) Bioactivity
Formulation Lyophilized from acetonitrile/TFA
Gene ID (Entrez) 7043
Molecular Weight (g/mol) M.W. (Observed): Dimeric TGF-beta 3 protein only, M.W. (Theroretical): 12.7 kDa (monomer), 25.4 kDa (dimer)
Quantity 500 μg
Source E. coli-derived human TGF-beta 3 protein
Storage Requirements Store lyophilized protein between -20°C to -80°C until the date of expiry. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias ARVD, FLJ16571, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3
Conjugate Unconjugated
Reconstitution Resuspend in 10 mM HCl at >100 μg/mL, prepare single use aliquots, add carrier protein if desired.
Protein TGF-beta 3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.