Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human TGF-beta RI/ALK-5 Isoform 1 Protein
SDP

Catalog No. NBP26198550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP26198550 50 μg
NB261985250 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP26198550 Supplier Novus Biologicals™ Supplier No. NBP26198550UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal hIgG-His-tag and corresponding to the amino acids 27-126 of Human TGF-beta RI/ALK-5 The Recombinant Human TGF-beta RI/ALK-5 Isoform 1 Protein is derived from Baculovirus. The Recombinant Human TGF-beta RI/ALK-5 Isoform 1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5 mg/ml
For Use With (Application) SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol.
Gene ID (Entrez) 7046
Molecular Weight (g/mol) 38 kDa
Quantity 50 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1μg of protein (determined by LAL method)
Gene Symbol TGFBR1
Cross Reactivity TGF-beta RI/ALK-5
Species Human
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.