Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human TNF-alpha (HEK293-expressed) Protein

Catalog No. p-7182948
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
50 μg
500 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Measured in a cytotoxicity assay using L-929 mouse fibroblast cells in the presence of the metabolic inhibitor actinomycin D. ED50 for this effect is 20 to 100 pg/mL.

Specifications

Accession Number P01375
For Use With (Application) Bioactivity
Gene ID (Entrez) 7124
Molecular Weight (g/mol) 17 kDa
Name APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Quantity 1 mg
Source Human embryonic kidney cell, HEK293-derived human TNF-alpha protein Val77-Leu233
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.