Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human TNF-alpha Protein
SDP

Catalog No. NBC118460 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118460 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118460 Supplier Novus Biologicals™ Supplier No. NBC118460
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

An un-tagged recombinant protein corresponding to the amino acids 77-233 of Human TNF-alpha The Recombinant Human TNF-alpha Protein is derived from E. coli. The Recombinant Human TNF-alpha Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Accession Number P01375
For Use With (Application) Bioactivity, SDS-PAGE
Formulation PBS (pH 7.4)
Gene ID (Entrez) 7124
Molecular Weight (g/mol) 17.5kDa
Name TNF-alpha Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Tumor Necrosis Factor-alpha Human, Recombinant, E.coli (NP_000585).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.