Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human TRAIL R2/TNFRSF10B Isoform 1 Protein
SDP

Catalog No. NBP26198350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP26198350 50 μg
NBP26198310 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP26198350 Supplier Novus Biologicals™ Supplier No. NBP26198350UG
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page, Bioactivity

A Biologically active recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 56-210 of Human TRAIL R2/TNFRSF10B The Recombinant Human TRAILR2/TNFRSF10B Isoform 1 Protein is derived from Baculovirus. The Recombinant Human TRAILR2/TNFRSF10B Isoform 1 Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration 1 mg/ml
For Use With (Application) Bioactivity, SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol.
Gene ID (Entrez) 8795
Molecular Weight (g/mol) 43.9 kDa
Quantity 50 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1μg of protein (determined by LAL method)
Gene Symbol TNFRSF10B
Cross Reactivity TRAIL R2/TNFRSF10B
Species Human
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.