Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Transthyretin/Prealbumin Protein
SDP

Catalog No. NBP137075 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP137075 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP137075 Supplier Novus Biologicals™ Supplier No. NBP137075
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 21-147 of Human Transthyretin/Prealbumin The Recombinant Human Transthyretin/Prealbumin Protein is derived from E. coli. The Recombinant Human Transthyretin/Prealbumin Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_000362
For Use With (Application) ELISA, Functional Assay, SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 7276
Molecular Weight (g/mol) 13.8kDa
Name Transthyretin/Prealbumin Protein
Purification Method IgG purified
Quantity 0.1 mg
Immunogen MGPTGTGESK CPLMVKVLDA VRGSPAINVA VHVFRKAADD TWEPFASGKT SESGELHGLT TEEEFVEGIY KVEIDTKSYW KALGISPFHE HAEVVFTAND SGPRRYTIAA LLSPYSYSTT AVVTNPKE
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Endotoxin Concentration <1 EU per 1ug of protein (determined by LAL method)
Cross Reactivity Human
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.