Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Recombinant Human Transthyretin/Prealbumin Y78F Variant, Monomer Protein
SDP

Catalog No. NB159067 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
2 x 100 μg Vials
5 x 100 μg Vials
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB159067 100 μg
NB159068 2 x 100 μg Vials
NB159069 5 x 100 μg Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB159067 Supplier Novus Biologicals Supplier No. NBP314787100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

For Use With (Application) In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot
Formulation PBS pH 7.4
Gene ID (Entrez) 7276
Quantity 100 μg
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias ATTR, HsT2651, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, thyroxine-binding prealbumin, transthyretin
Gene Symbol TTR
Research Category Cancer
Purity or Quality Grade >95%, by SDS-PAGE
Protein Transthyretin/Prealbumin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.