Learn More
Description
Specifications
Specifications
| Accession Number | P07101-2 |
| For Use With (Application) | Immunohistochemistry-Paraffin, SDS-PAGE, Western Blot |
| Formulation | 300 mM NaCl, 2.7 mM KCl, 4.3 mM Na2HPO4, 1.47 mM KH2PO4, 5% glycerol, 1 mM DTT, pH 7.4 |
| Gene ID (Entrez) | 7054 |
| Molecular Weight (g/mol) | M.W. theroretical: 60 kDa |
| Name | Tyrosine Hydroxylase Protein |
| Quantity | 20 μg |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Research Category | Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Prostate Cancer |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
