Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Ubiquitin C Protein
SDP

Catalog No. NBC118439 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118439 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118439 Supplier Novus Biologicals™ Supplier No. NBC118439
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-76 of Human Ubiquitin C The Recombinant Human Ubiquitin C Protein is derived from E. coli. The Recombinant Human Ubiquitin C Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number CAA44911
For Use With (Application) ELISA, SDS-PAGE
Formulation 50mM HEPES buffer (pH7.5) 150mM NaCl, 1mM DTT, 10% glycerol
Gene ID (Entrez) 7316
Molecular Weight (g/mol) 8.6kDa
Name Ubiquitin C Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MQIFVKTLTGKTITLEVEPSDTIENVKAKIQDKEGIPPDQQRLIFAGKQLEDGRTLSDYNIQKESTLHLVLRLRGG
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.