Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Universal Type I IFN Protein
Click to view available options
Quantity:
10 μg
50 μg
Description
Measured in anti-viral assays using HeLa human cervical epithelial carcinoma cells infected with encephalomyocarditis (EMC) virus. Meager, A. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 129. The ED50 for this effect is 6.00-60.0 pg/mL.
Specifications
Specifications
Accession Number | P01563.1 |
For Use With (Application) | Bioactivity |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
Gene ID (Entrez) | 3440 |
Molecular Weight (g/mol) | MolecularWeight-observed: 16-22 kDa, under reducing conditions.; MolecularWeight-theroretical: 19 kDa |
Quantity | 10 μg |
Source | Human embryonic kidney cell, HEK293-derived human IFN-alpha protein Human IFNA-2 A (Cys24-Gln85) (N-terminus) Accession # P01563.1 Human IFNA-1 B (Ile87-Glu189) (N-terminus) Accession # CAA23799.1 (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
Gene Alias | IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction