Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human VEGF 121 Protein
SDP

Catalog No. NB398122 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398122 10 μg
NB398123 100 μg
NB398121 1 mg
NB398120 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. NB398122 Supplier Novus Biologicals™ Supplier No. NBP30710310UG
Add to Cart
Add to Cart

Recombinant Protein

Fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human umbilical vein endothelial cells (HUVEC) is between 0.2-0.4 ng/mL.

Specifications

Accession Number P15692-9
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in PBS, (pH 7.4)
Gene ID (Entrez) 7422
Quantity 10 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 0.1 EU/μg of VEGF 121 AS determined by LAL method.
Gene Alias MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Gene Symbol VEGFA
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Briefly centrifuge vial prior to opening to bring the contents to the bottom. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to 0.1-1.0 mg/mL Stock solutions should be apportioned into working aliquots and stored (at -20°C.)
Form Purified
Research Category Angiogenesis, Biologically Active Proteins
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein VEGF 121
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.