Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human ZBTB6 GST (N-Term) Protein

Click to view available options
Quantity:
2 μg
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Specifications
For Use With (Application) | Western Blot, ELISA, SDS-PAGE, Protein Array, Immunoaffinity Purification |
Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
Gene ID (Entrez) | 10773 |
Molecular Weight (g/mol) | 74.6 kDa |
Quantity | 2 μg |
Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
Gene Alias | zinc finger and BTB domain containing 6, zinc finger and BTB domain-containing protein 6, Zinc finger protein 482ZIDZinc finger protein with interaction domain, ZNF482 |
Gene Symbol | ZBTB6 |
Research Category | Cancer, Cell Cycle and Replication |
Purity or Quality Grade | >80% by SDS Page and Coomassie blue staining |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction