Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Insect TDO2 His Protein
SDP

Catalog No. NB171729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171729 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171729 Supplier Novus Biologicals™ Supplier No. NBP318168
This item is not returnable. View return policy

Recombinant Protein

Specific activity towards L-Trp: 87.0 umol/hr per mg. TDO catalyzes the oxidative opening of the L-tryptophan indole moiety to yield N-formylkynurenine (NFK).

Specifications

Accession Number O77457
For Use With (Application) Enzyme Activity
Formulation 50mM sodium phosphate pH 8.0, 0.15M NaCl, 0.2M imidazole and 10% glycerol
Gene ID (Entrez) 6999
Molecular Weight (g/mol) M.W. Theoretical: 47.9 kDa
Quantity 50 μg
Storage Requirements Store at -20°C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Gene Alias TDOTO, TPH2EC 1.13.11.11, TRPOtryptophanase, Tryptamin 2,3-dioxygenase, tryptophan 2,3-dioxygenase, Tryptophan oxygenase, Tryptophan pyrrolase, Tryptophanase
Gene Symbol TDO2
Research Category Immunology, Lipid and Metabolism, Neuroscience
Purity or Quality Grade >98%, by SDS-PAGE
Protein TDO2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.