Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Monkeypox Virus A33R His (N-Term) GST (N-Term) Protein

Click to view available options
Quantity:
100 μg
Specifications
Specifications
For Use With (Application) | SDS-PAGE |
Formulation | Lyophilized from sterile 50mM Tris, 150mM NaCl, 1mM EDTA, 10% Glycerol, pH 8.0. Normally 5%-8% trehalose, mannitol and 0.01% Tween80 are added as protectants before lyophilization. Please contact technical support. |
Molecular Weight (g/mol) | M.W. Theoretical: 44.07 kDa |
Quantity | 100 μg |
Storage Requirements | Store at -20 to -80C. Avoid freeze-thaw cycles. |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | MPXV-A33R |
Reconstitution | Reconstitution instructions are lot-specific, please contact technical support |
Purity or Quality Grade | >85%, by SDS-PAGE |
Protein | Monkeypox Virus A33R |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction