Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Active JNK1 Protein
Click to view available options
Quantity:
10 μg
Description
The specific activity of JNK1 was determined to be 104 nmol/min/mg using an ATF2 substrate and was equivalent to 122 nmol/min/mg as per radiometric assay.
Specifications
Specifications
Accession Number | NP_002741 |
For Use With (Application) | Bioactivity |
Formulation | Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol. |
Gene ID (Entrez) | 5599 |
Molecular Weight (g/mol) | M.W. (Observed): 70 kDa |
Quantity | 10 μg |
Source | Spodoptera frugiperda, Sf 9 (baculovirus)-derived mouse JNK1 protein |
Storage Requirements | This product is stable at <= -70°C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store (at recommended temperature. Avoid repeated freeze-thaw cycles.) |
Gene Symbol | Mapk9 |
Conjugate | Unconjugated |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction