Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Adiponectin/Acrp30 Protein

Catalog No. 5095AC050
Click to view available options
:
50ug; Carrier Free
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Adiponectin/Acrp30 Protein is derived from NS0. The Recombinant Mouse Adiponectin/Acrp30 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Supplied as a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 11450
Molecular Weight (g/mol) 25.8 kDa
Name Adiponectin/Acrp30
Quantity 50μg
Source Mouse myeloma cell line,NS0-derived mouse Adiponectin/Acrp30 protein Glu18-Asn247,with a C-terminal 6-His tag
Storage Requirements Do not freeze. 6 months from date of receipt, 2 to 8°C as supplied.
Structural Form Oligomer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.