Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse ADK Protein
SDP

Catalog No. NBP26119350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP26119350 50 μg
NBP26119310 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP26119350 Supplier Novus Biologicals™ Supplier No. NBP26119350UG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-361 of Mouse ADK The Recombinant Mouse ADK Protein is derived from E. coli. The Recombinant Mouse ADK Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 1 mg/ml
For Use With (Application) Bioactivity, Functional Study, SDS-PAGE
Formulation PBS (pH 7.4) containing 10% glycerol.
Gene ID (Entrez) 132
Molecular Weight (g/mol) 42.5 kDa
Quantity 50 μg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1 μg of protein (determined by LAL method)
Gene Symbol ADK
Cross Reactivity ADK
Species Mouse
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.