Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse AHSA2 His Protein
SDP

Catalog No. NB172000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172000 50 μg
NB171999 200 μg
NB171998 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB172000 Supplier Novus Biologicals™ Supplier No. NBP31832650UG
This item is not returnable. View return policy

Recombinant Protein

Specifications

For Use With (Application) SDS-PAGE, Western Blot
Formulation 25mM Trif buffer pH7.2, 150mM NaCl, 0.1mM DTT, 50% glycerol
Gene ID (Entrez) 130872
Molecular Weight (g/mol) M.W. Theoretical: 38 kDa
Quantity 50 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias activator of 90 kDa heat shock protein ATPase homolog 2, AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast), DKFZp564C236, FLJ34679, FLJ41715, Hch1
Gene Symbol AHSA2P
Research Category Cancer, Cellular Signaling, Chaperone Mediated Autophagy (CMA)
Purity or Quality Grade >85%
Protein AHSA2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.