Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse alpha-L-Iduronidase/IDUA Protein

Catalog No. 9348GH010
Click to view available options
Quantity:
10 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave a fluorogenic substrate, 4-Methylumbelliferyl alpha-L-iduronide. The specific activity is >7,500 pmol/min/ug, as measured under the described conditions.

Order Info

Ships at ambient temperature.

Specifications

Accession Number P48441
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Sodium Acetate, NaCl and Glycerol.
Gene ID (Entrez) 3425
Molecular Weight (g/mol) M.W. (Observed): 83-95 kDa, reducing conditions; M.W. (theoretical): 70 kDa
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived mouse alpha-L-Iduronidase/IDUA protein Glu17-Ser634, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias alphaLIduronidase, IDA, IDUA, MPS1, MPSI
Biological Activity >7,500pmol/min/μg
Product Type Recombinant protein
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein alpha-L-Iduronidase/IDUA
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.