Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse alpha-Synuclein (Control) Protein, Pre-formed Fibrils
SDP

Catalog No. NB261596100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
2 x 100 μg Vials
5 x 100 μg Vials
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB261596100 100 μg
NB398300 2 x 100 μg Vials
NB398299 5 x 100 μg Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB261596100 Supplier Novus Biologicals™ Supplier No. NBP261596100UG
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results.

Active Mouse Recombinant Alpha Synuclein Protein Aggregate (pre-formed fibrils, Type 1). NCBI Accession #: NP_001035916.1. The Recombinant Mouse alpha-Synuclein (Control) Protein is derived from E. coli. The Recombinant Mouse alpha-Synuclein (Control) Protein has been validated for the following applications: Western Blot, In vitro assay, In vivo assay, SDS-Page, Bioactivity.

Specifications

Concentration 2 mg/ml
For Use With (Application) Bioactivity, In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot
Formulation PBS
Gene ID (Entrez) 6622
Molecular Weight (g/mol) 14.46 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at -80°C in the dark. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol SNCA
Cross Reactivity alpha-Synuclein
Species Mouse
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.