Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse B7-1/CD80 Protein
SDP

Catalog No. NBP226578 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226578 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226578 Supplier Novus Biologicals™ Supplier No. NBP226578
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page, Cell depletion

A recombinant protein corresponding to CD80.

Amino Acid Sequence: MGHTRRQGTSPSKCPYLNFFQLLVLAGLSHFCSGVIHVTKEVKEVATL SCGHNVSVEELAQTRIYWQKEKKMVLTMMSGDMNIWPEYKNRTIFDI TNNLSIVILALRPSDEGTYECVVLKYEKDAFKREHLAEVTLSVKADFPT PSISDFEIPTSNIRRIICSTSGGFPEPHLSWLENGEELNAINTTVSQDPETEL YAVSSKLDFNMTTNHSFMCLIKYGHLRVNQTFNWNTTKQEHFPDDKTHT CPPCPAPELLGGPSVFLFPPKPKDTLMISRTPEVTCVVVDVSHEDPEVKFNWYV DGVEVHNAKTKPREEQYNSTYRVVSVLTVLHQDWLNGKEYKCKV SNKALPAPIEKTISKAKGQPREPQVYTLPPSRDELTKNQVSLTCLVKGFYPSDI AVEWESNGQPENNYKTTPPVLDSDGSFFLYSKLTVDKSRWQQGNVFSCSVM HEALHNHYTQKSLSLSPGK The Recombinant Mouse B7-1/CD80 Protein has been validated for the following applications: SDS-Page, Cell depletion.

Specifications

Gene ID (Entrez) 941
Name B7-1/CD80 Protein
Purification Method Protein A purified
Quantity 0.05 mg
Source Baculovirus
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.