Learn More
Description
A recombinant protein with a His-Tag and corresponding to the amino acids 29-248aa of Mouse B7-H3.
Source: Baculovirus
Amino Acid Sequence:
VEVQVSEDPV VALVDTDATL RCSFSPEPGF SLAQLNLIWQ LTDTKQLVHS FTEGRDQGSA YSNRTALFPD LLVQGNASLR LQRVRVTDEG SYTCFVSIQD FDSAAVSLQV AAPYSKPSMT LEPNKDLRPG NMVTITCSSY QGYPEAEVFW KDGQGVPLTG NVTTSQMANE RGLFDVHSVL RVVLGANGTY SCLVRNPVLQ QDAHGSVTIT GQPLTFPPEA HHHHHH>
Specifications
Specifications
| Concentration | 0.25 mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | PBS (pH 7.4), 10% glycerol |
| Gene ID (Entrez) | 80381 |
| Molecular Weight (g/mol) | 24.7 kDa |
| Quantity | 0.05 mg |
| Source | Baculovirus |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule |
| Gene Symbol | CD276 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
