Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse CAMK2N1 Protein
SDP

Catalog No. NBP25957710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25957710 10 μg
NBP25957750 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25957710 Supplier Novus Biologicals™ Supplier No. NBP25957710UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-78 of Mouse CAMK2N1 The Recombinant Mouse CAMK2N1 Protein is derived from E. coli. The Recombinant Mouse CAMK2N1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris-HCl (pH8.5) containing 1mM DTT, 0.1M NaCl, 10% glycerol
Gene ID (Entrez) 55450
Molecular Weight (g/mol) 10.9 kDa
Name Recombinant Mouse CAMK2N1 Protein
Purification Method >90%, by SDS-PAGE
Quantity 10 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias Calcium/Calmodulin Dependent Protein Kinase II Inhibitor 1, Calcium/Calmodulin-Dependent Protein Kinase II Inhibitor 1, CaMKII Inhibitory Protein Alpha, CaMKIINalpha, CaMKIIN-Alpha, PRO1489
Gene Symbol VAMK2N1
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Mouse
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.