Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse Cathepsin D Protein
SDP

Catalog No. NBP2595245 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
5 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2595245 5 μg
NBP25952420 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP2595245 Supplier Novus Biologicals™ Supplier No. NBP2595245UG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 21-410 of Mouse Cathepsin D The Recombinant Mouse Cathepsin D Protein is derived from E. coli. The Recombinant Mouse Cathepsin D Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 0.25mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid. PBS (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 1509
Molecular Weight (g/mol) 44 kDa
Name Recombinant Mouse Cathepsin D Protein
Purification Method >90%, by SDS-PAGE
Quantity 5 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1 microgram of protein (determined by LAL method)
Gene Alias cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10
Gene Symbol CTSD
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Mouse
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.