Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Cathepsin E Protein
Click to view available options
Quantity:
10 μg
Description
Measured by its ability to cleave the fluorogenic peptide substrate, Mca-PLGL-Dpa-AR-NH2 (Catalog # ES001 ). The specific activity is >8,000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
Accession Number | P70269 |
For Use With (Application) | Enzyme Activity |
Formulation | Supplied as a 0.2 μm filtered solution in MES and NaCl. |
Gene ID (Entrez) | 1510 |
Molecular Weight (g/mol) | M.W. (Observed): 53 kDa, reducing conditions; M.W. (theoretical): 42 & 38 kDa |
Name | Cathepsin E |
Quantity | 10 μg |
Source | Mouse myeloma cell line, NS0-derived mouse Cathepsin E protein Gln19-Pro397, with a C-terminal 10-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
Structural Form | Disulfide-linked homodimer |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction