Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse CD24 Fc Protein
SDP

Catalog No. NB171711 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171711 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171711 Supplier Novus Biologicals™ Supplier No. NBP318150
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number P25063
Concentration LYOPH
For Use With (Application) SDS-PAGE
Formulation Lyophilized from PBS
Gene ID (Entrez) 100133941
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.06 EU/μg purified protein (LAL test; Lonza).
Gene Alias CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen
Gene Symbol CD24
Reconstitution Reconstitute with 50 ul sterile water. Add 1X PBS to the desired protein concentration (1mg/ml after reconstitution)
Research Category B Cell Development and Differentiation Markers, Cancer, Neuronal Cell Markers, Stem Cell Markers
Purity or Quality Grade >90% SDS-PAGE
Protein CD24
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.