Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse CD27 Ligand/TNFSF7 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Description
Measured by its ability to stimulate mouse T cell proliferation in the presence of anti-CD3. The ED50 for this effect is 1.25-5.0 μg/mL.
Specifications
Specifications
| Accession Number | Q05A52 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 970 |
| Molecular Weight (g/mol) | M.W. (Observed): 30 kDa, reducing conditions; M.W. (theoretical): 19.2 kDa |
| Name | CD27 Ligand/TNFSF7 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse CD27 Ligand/TNFSF7 protein MHHHHHHHHHH GGGSGGGSGGGS IEGR Mouse CD27 Ligand (Gln47 - Pro195) (N-terminus) Accession # Q05A52 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction