Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse Darcin Protein
SDP

Catalog No. NB171712 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171712 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171712 Supplier Novus Biologicals™ Supplier No. NBP318151
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number Q5FW60
Concentration LYOPH
For Use With (Application) SDS-PAGE
Formulation Lyophilized from PBS
Gene ID (Entrez) 381530
Molecular Weight (g/mol) M.W. Theoretical: 17 kDa
Quantity 10 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.1 EU/μg purified protein (LAL test; Lonza).
Gene Alias Major urinary protein 20, Major urinary protein 24, Mup20
Gene Symbol Mup20
Reconstitution Reconstitute with 100 ul sterile distilled water (0.1mg/ml after reconstitution)
Research Category Immunology
Purity or Quality Grade >95% pure by SDS-PAGE
Protein Darcin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.