Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse EGF Isoform 1 Protein
SDP

Catalog No. NBP2619815 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
5 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2619815 5 μg
NBP26198120 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP2619815 Supplier Novus Biologicals™ Supplier No. NBP2619815UG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page, Bioactivity

A Biologically active recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 977-1029 of Mouse EGF The Recombinant Mouse EGF Isoform 1 Protein is derived from E. coli. The Recombinant Mouse EGF Isoform 1 Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration 0.25 mg/ml
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris-HCl(pH8.0) containing 0.1M NaCl, 2mM DTT, 10% glycerol
Gene ID (Entrez) 1950
Molecular Weight (g/mol) 8.6 kDa
Quantity 5 μg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 0.2 EU per 1μg of protein (determined by LAL method)
Gene Symbol EGF
Cross Reactivity EGF
Species Mouse
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.