Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Frizzled-5 Fc Chimera Protein

Catalog No. 9060FZ050
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Frizzled-5 Fc Chimera Protein is derived from NS0. The Recombinant Mouse Frizzled-5 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9EQD0
For Use With (Application) Bioactivity
Gene ID (Entrez) 7855
Molecular Weight (g/mol) Observed M.W.: 54-64kDa, reducing conditions, Theoretical M.W.: 43kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse Frizzled-5 protein, (N-terminus) Mouse Frizzled-5 (Ala27-Pro167) Accession &Num; Q9EQD0 IEGRMD Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias C2orf31, chromosome 2 open reading frame 31, DKFZP434E2135, frizzled (Drosophila) homolog 5, frizzled homolog 5 (Drosophila), frizzled-5, Frizzled5, Frz-5, FZ-5, FZD5, FzE5, HFZ5, MGC129692, seven-transmembrane receptor frizzled-5, Wnt receptor
Common Name Frizzled-5
Gene Symbol Fzd5
Biological Activity Measured by its ability to inhibit Wnt-3a-induced alkaline phosphatase production by MC3T3-E1 mouse preosteoblast cells. The ED50 for this effect is typically 10-60ng/mL in the presence of 5ng/mL of Recombinant Mouse Wnt-3a.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Mouse Frizzled-5 (Ala27-Pro167) Accession # Q9EQD0, N-terminus; IEGRMD; Mouse IgG2a (Glu98-Lys330), C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.