Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Growth Hormone R (GHR) Fc Chimera
Click to view available options
Quantity:
50 μg
Description
The Recombinant Mouse Growth Hormone R (GHR) Fc Chimera is derived from NS0. The Recombinant Mouse Growth Hormone R (GHR) Fc Chimera has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
Gene ID (Entrez) | 14600 |
Molecular Weight (g/mol) | 55.4 kDa (monomer) |
Name | Growth Hormone R Fc Chimera |
Quantity | 50 μg |
Source | Mouse myeloma cell line,NS0-derived mouse Growth Hormone R/GHR protein, (N-terminus) Mouse GHR (Thr25-Gln273) & (Ile98-Gln273) Accession &Num; Q3UP14 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked homodimer |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction