Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Hepsin Protein

Catalog No. 9156SE010
Click to view available options
Quantity:
10 μg
This item is not returnable. View return policy

Catalog No. 91-56S-E010

Supplier: R&D Systems™ 9156SE010

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave tert-butoxycarbonyl-Gln-Arg-Arg-7-amino-4-methylcoumarin (Boc-QRR-AMC). The specific activity is >8,000 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number O35453-2
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Sodium Acetate and NaCl.
Gene ID (Entrez) 3249
Molecular Weight (g/mol) M.W. (Observed): 41-47 kDa, reducing conditions; M.W. (theoretical): 42 kDa
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived mouse Hepsin protein Gln44-Pro416 (Asp160Glu & Arg161Lys), with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias EC 3.4.21, EC 3.4.21.106, hepsin, HPN, serine protease hepsin, serine protease hepsin catalytic chain, TMPRSS1 TMPRSS1serine protease hepsin non-catalytic chain, Transmembrane protease serine 1, transmembrane protease, serine 1
Common Name Hepsin
Biological Activity Measured by its ability to cleave tert-butoxycarbonyl-Gln-Arg-Arg-7-amino-4-methylcoumarin (Boc-QRR-AMC). The specific activity is >8,000 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Gln44-Pro416 (Asp160Glu & Arg161Lys), with a C-terminal 10-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Hepsin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.