Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse Histone Deacetylase 8/HDAC8 His Protein
SDP

Catalog No. NBP25951810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25951810 10 μg
NBP25951850 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25951810 Supplier Novus Biologicals™ Supplier No. NBP25951810UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-377 of Mouse Histone Deacetylase 8/HDAC8 The Recombinant Mouse Histone Deacetylase 8/HDAC8 Protein is derived from E. coli. The Recombinant Mouse Histone Deacetylase 8/HDAC8 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. PBS (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 55869
Molecular Weight (g/mol) 42.5 kDa
Name Recombinant Human Histone Deacetylase 8/HDAC8 Protein
Purification Method >90%, by SDS-PAGE
Quantity 10 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1 microgram of protein (determined by LAL method)
Gene Alias EC 3.5.1.98, HD8, HDACL1, histone deacetylase 8, histone deacetylase-like 1, RPD3
Gene Symbol HDAC8
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.