Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IFN-alpha 1 Protein

Catalog No. p-7180211
Click to view available options
Quantity:
10 μg
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Human embryonic kidney cell, HEK293-derived mouse IFN-alpha 1 protein

  • The interferons (IFN) are a family of cytokines with potent antiviral, antiproliferative and immunomodulatory properties, and classified based on their binding specificity to cell surface receptors
  • IFN-alpha 1 Protein (aka IFNA1 or IFNalpha 1)

Specifications

Accession Number P01572
For Use With (Application) Bioactivity
Gene ID (Entrez) 3439
Molecular Weight (g/mol) 19 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 10 μg
Source Human embryonic kidney cell, HEK293-derived mouse IFN-alpha 1 protein Cys24-Lys189
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after opening. 3 months, -20 to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.