Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse IFN-alpha 12 Protein
Click to view available options
Quantity:
10 μg
50 μg
Description
Vogel, S.N. et al. (1982) Infect. Immunol. 38:681. The ED50 for this effect is 10-100 pg/mL.
Specifications
Specifications
Accession Number | Q80SS5.1 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
Molecular Weight (g/mol) | Observed: 18-25 kDa, under reducing conditions; Theroretical: 19.2 kDa |
Quantity | 10 μg |
Source | Human embryonic kidney cell, HEK293-derived mouse IFN-alpha 12 protein Cys24-Glu189 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
Conjugate | Unconjugated |
Reconstitution | Reconstitute at 100 μg/mL in PBS. |
Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction