Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse IL-27 Protein
SDP

Catalog No. N276310100U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N276310100U 100 μg
N276310500U 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N276310100U Supplier Novus Biologicals™ Supplier No. NBP276310100UG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page, Bioactivity

An un-tagged bioactive recombinant protein corresponding to Mouse IL-27

The Recombinant Mouse IL-27 Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration Lyoph
For Use With (Application) SDS-PAGE, Bioactivity
Gene ID (Entrez) 246778
Molecular Weight (g/mol) 23.7 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration Acceptance criteria for endotoxin levels are <1 EUs/ug protein (determined by Kinetic LAL Method).
Gene Symbol IL27
Specific Reactivity IL-27
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Reconstitution Sterile water at 0.1 mg/mL
Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium bicarbonate, pH 8.5
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.