Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse IL-38/IL-1F10 FLAG Protein
SDP

Catalog No. NB171716 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171716 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171716 Supplier Novus Biologicals™ Supplier No. NBP318155
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number Q8R459
Concentration LYOPH
For Use With (Application) SDS-PAGE
Formulation Lyophilized from PBS + 0.5% Trehalose
Gene ID (Entrez) 84639
Molecular Weight (g/mol) M.W. Theoretical: 20 kDa
Quantity 10 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.01 EU/μg purified protein (LAL test; Lonza).
Gene Alias interleukin 1 family, member 10 (theta), UNQ6119/PRO20041
Gene Symbol IL1F10
Reconstitution Reconstitute with 100 ul sterile water (0.1mg/ml after reconstitution)
Research Category Immunology
Purity or Quality Grade >95% pure by SDS-PAGE
Protein IL-38/IL-1F10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.