Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse IL-3R alpha/CD123 hIgG-His Protein
SDP

Catalog No. NB172125 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172125 50 μg
NB172124 250 μg
NB172123 20 μg
NB172122 10 μg
NB172121 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. NB172125 Supplier Novus Biologicals™ Supplier No. NBP31835950UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

Concentration 0.25 mg/ml
For Use With (Application) Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin, SDS-PAGE, Western Blot
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 30% glycerol
Gene ID (Entrez) 3563
Molecular Weight (g/mol) M.W. Theoretical: 61.5 kDa
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <1 EU per 1ug of protein (determined by LAL method)
Gene Alias CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Gene Symbol IL3RA
Research Category B Cell Development and Differentiation Markers, Cytokine Research, Oncogenes, Phospho Specific, Protein Kinase
Purity or Quality Grade >90%, by SDS-PAGE
Protein IL-3R alpha/CD123
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.