Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
R&D Systems™ Recombinant Mouse Nodal Protein
                    Click to view available options
                
                
                    
                            
                                Formulation:
                            
                        
                        
                            
                                Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
                            
                        
                            
                                Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
                            
                        
                    Description
The Recombinant Mouse Nodal Protein is derived from E. coli. The Recombinant Mouse Nodal Protein has been validated for the following applications: Bioactivity.
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
                
            Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | 
| Gene ID (Entrez) | 18119 | 
| Molecular Weight (g/mol) | 13 kDa (monomer) | 
| Name | Nodal | 
| Quantity | 25 μg | 
| Source | E. coli-derived mouse Nodal protein His245-Leu354,with an N-terminal Met | 
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | 
| Structural Form | Disulfide-linked homodimer | 
| Conjugate | Unconjugated | 
| Recombinant | Recombinant | 
| Show More | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            