Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse Park7/DJ-1 His Protein
SDP

Catalog No. NBP259523X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
NBP259523X 100 μg
NBP25952305 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. NBP259523X Fournisseur Novus Biologicals™ Code fournisseur NBP259523100UG
Ajouter au panier
Ajouter au panier

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-189 of Mouse Park7/DJ-1 The Recombinant Mouse Park7/DJ-1 Protein is derived from E. coli. The Recombinant Mouse Park7/DJ-1 Protein has been validated for the following applications: SDS-Page.

Spécifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. PBS (pH 7.4) containing 20% glycerol,1mM DTT
Gene ID (Entrez) 11315
Molecular Weight (g/mol) 22.4 kDa
Name Recombinant Mouse Park7/DJ-1 Protein
Purification Method >90%, by SDS-PAGE
Quantity 100 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1
Gene Symbol PARK7
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Mouse
Recombinant Recombinant
Afficher plus de résultats Afficher moins de résultats

For Research Use Only.

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.