Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse PDGF-B Protein
SDP

Catalog No. NB171777 Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

3T3 cell proliferation, >5.0 x 10^5 units/mg (ED50 acceptance criteria are <2 ng/mL).

Specifications

For Use With (Application) Bioactivity, SDS-PAGE
Formulation Lyophilized from a 0.2 micron filtered solution, 10 mM sodium citrate, pH 3.0
Gene ID (Entrez) 5155
Quantity 500 μg
Storage Requirements Store at -20 to -70C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20 to -70C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Acceptance criteria for endotoxin levels are <1 EUs/μg protein (determined by Kinetic LAL Method).
Gene Alias B chain, Becaplermin, FLJ12858, PDGF subunit B, PDGF2, PDGF-2, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SSV
Gene Symbol PDGFB
Reconstitution Sterile water at 0.1 mg/mL
Research Category Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Markers
Purity or Quality Grade >95%, by SDS-PAGE
Protein PDGF-B
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.