Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse PDGFA Protein

Click to view available options
Quantity:
1 mg
10 μg
100 μg
Description
3T3 cell proliferation, >1.0 x 10^5 units/mg (ED50 acceptance criteria are <10 ng/mL).
Specifications
Specifications
For Use With (Application) | Bioactivity, SDS-PAGE |
Formulation | Lyophilized from a 0.2 micron filtered solution, 0.1% Trifluoroacetic Acid (TFA) |
Gene ID (Entrez) | 5154 |
Quantity | 1 mg |
Storage Requirements | Store at -20 to -70C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20 to -70C for long term storage. Avoid repeated freeze-thaw cycles. |
Endotoxin Concentration | Acceptance criteria for endotoxin levels are <1 EUs/μg protein (determined by Kinetic LAL Method). |
Gene Alias | PDGF-1, PDGF1PDGF A-chain, PDGF-A, Platelet-derived growth factor A chain, platelet-derived growth factor alpha chain, platelet-derived growth factor alpha isoform 2 preproprotein, platelet-derived growth factor alpha polypeptidePDGF subunit A, platelet-derived growth factor subunit A |
Gene Symbol | PDGFA |
Reconstitution | Sterile water at 0.1 mg/mL |
Research Category | Cell Cycle and Replication |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction