Learn More
Description
Specifications
Specifications
| Accession Number | NP_035229, NP_035229 |
| Concentration | 0.5 mg/ml |
| Formulation | 20 mM Tris-HCl buffer (pH 8.5), 0.2 M NaCl, 1 mM DTT, 30% glycerol |
| Gene ID (Entrez) | 5315 |
| Molecular Weight (g/mol) | Theroretical: 60.2 kDa |
| Quantity | 20 μg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Gene Symbol | PKM |
| Purity or Quality Grade | >85%, by SDS-PAGE |
| Protein | PKM2 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
