Learn More
Novus Biologicals™ Recombinant Mouse Prostatic Acid Phosphatase/ACPP His (C-Term) Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Concentration | 0.5 mg/mL |
| For Use With (Application) | Enzyme Activity, Functional Assay, SDS-PAGE |
| Formulation | PBS (pH 7.4) and 10% glycerol |
| Gene ID (Entrez) | 55 |
| Molecular Weight (g/mol) | 41.3 kDa |
| Quantity | 50 μg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of protein (determined by LAL method) |
| Gene Alias | acid phosphatase, prostate, ACP3, ACP-3, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase |
| Gene Symbol | ACP3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.