Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse R-Spondin 1 (CHO-expressed) Protein

Catalog No. 7150RS250CF
Click to view available options
Quantity:
250 μg
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse R-Spondin 1 (CHO-expressed) Protein is derived from CHO. The Recombinant Mouse R-Spondin 1 (CHO-expressed) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_619624
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 284654
Molecular Weight (g/mol) 27 kDa
Quantity 250 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse R-Spondin 1 protein Met1-Gln265
Storage Requirements Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
Regulatory Status RUO
Endotoxin Concentration <0.01 EU per 1 μg of the protein by the LAL method.
Gene Symbol Rspo1
Conjugate Unconjugated
Cross Reactivity R-Spondin 1
Species Mouse
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.