Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse RalA/RalB Protein
SDP

Catalog No. NB261199100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB261199100 100 μg
NBP26119905 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB261199100 Supplier Novus Biologicals™ Supplier No. NBP261199100UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids1-203 of MouseRalA/RalB The Recombinant Mouse RalA/RalB Protein is derived from E. coli. The Recombinant Mouse RalA/RalB Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5 mg/ml
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4) containing 10% glycerol.
Gene ID (Entrez) 5898
Molecular Weight (g/mol) 25.7 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol RALA
Cross Reactivity RalA/RalB
Species Mouse
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.