Learn More
Description
- Recombinant protein for Mouse RELM beta
- The Recombinant Mouse RELM beta Protein is derived from E. coli.
- The Recombinant Mouse RELM beta Protein has been validated for the following applications: SDS-Page.
- Amino Acid Sequence:(Q8K425)
Specifications
Specifications
| Concentration | Lyoph |
| For Use With (Application) | SDS-PAGE |
| Gene ID (Entrez) | 84666 |
| Molecular Weight (g/mol) | 8.893 kDa |
| Purification Method | >97%, by SDS-PAGE |
| Quantity | 25 μg |
| Source | E. coli |
| Storage Requirements | Store at -20 to -80°C. Avoid freeze-thaw cycles. |
| Gene Symbol | RETNLB |
| Reconstitution | Reconstitute the lyophilized product with sterile H2O to a concentration of 0.1 mg/mL, which can be further diluted into other aqueous solutions. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
