Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse SCF/c-kit Ligand Protein
SDP

Catalog No. NB171765 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171765 1 mg
NB171764 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB171765 Supplier Novus Biologicals™ Supplier No. NBP3181851MG
This item is not returnable. View return policy

Recombinant Protein

TF-1 cell proliferation, >5.0 x 10^4 units/mg (ED50 acceptance criteria are <20 ng/mL).

Specifications

For Use With (Application) Bioactivity, Immunohistochemistry, Immunohistochemistry-Paraffin, SDS-PAGE, Western Blot
Formulation Lyophilized from a 0.2 micron filtered solution, 20 mM sodium phosphate, pH 6.5
Gene ID (Entrez) 4254
Quantity 1 mg
Storage Requirements Store at -20 to -70C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20 to -70C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Acceptance criteria for endotoxin levels are <1 EUs/μg protein (determined by Kinetic LAL Method).
Gene Alias DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor
Gene Symbol KITLG
Reconstitution Sterile water at 0.1 mg/mL
Research Category Biologically Active Proteins, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
Purity or Quality Grade >95%, by SDS-PAGE
Protein SCF/c-kit Ligand
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.