Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse STI1 His Protein
SDP

Catalog No. NBP259538X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259538X 100 μg
NBP25953805 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP259538X Supplier Novus Biologicals™ Supplier No. NBP259538100UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-543 of Mouse STI1 The Recombinant Mouse STI1 Protein is derived from E. coli. The Recombinant Mouse STI1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris-HCl(pH8.0) containing 0.1M NaCl, 1mM DTT, 10% glycerol
Gene ID (Entrez) 10963
Molecular Weight (g/mol) 64.7 kDa
Name Recombinant Mouse STI1 Protein
Purification Method >90%, by SDS-PAGE
Quantity 100 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias Hop, HOPSTI1L, Hsc70/Hsp90-organizing protein, Hsp70/Hsp90-organizing protein, IEF-SSP-3521, NY-REN-11 antigen, Renal carcinoma antigen NY-REN-11, STI1P60, stress-induced-phosphoprotein 1, Transformation-sensitive protein IEF SSP 3521
Gene Symbol STIP1
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Mouse
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.