Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse Testican 3/SPOCK3 His Protein
SDP

Catalog No. NBP250571E Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250571E 20 μg
NBP250571D 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP250571E Supplier Novus Biologicals™ Supplier No. NBP25057120UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 22-436 of Mouse Testican 3/SPOCK3 The Recombinant Mouse Testican 3/SPOCK3 Protein is derived from Baculovirus. The Recombinant Mouse Testican 3/SPOCK3 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation In PBS (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 50859
Molecular Weight (g/mol) TMW: 47.9kDa
Quantity 20 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −70°C long term. Avoid freeze-thaw cycles.
Gene Alias CWCV, and Kazal-like domains proteoglycan 3, HSAJ1454, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3, TES-3, testican 3, TICN3
Common Name Testican 3/SPOCK3
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.